Skip to Content
Merck
All Photos(1)

Key Documents

S3125

Sigma-Aldrich

Bis(2-ethylhexyl) sebacate

~95%

Synonym(s):

Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
426.67
Beilstein:
1806504
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

~95%

refractive index

n20/D 1.450 (lit.)

bp

212 °C/1 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin R Wilson et al.
Physical chemistry chemical physics : PCCP, 14(4), 1468-1479 (2011-12-14)
The heterogeneous reactions of hydroxyl radicals (OH) with squalane and bis(2-ethylhexyl) sebacate (BES) particles are used as model systems to examine how distributions of reaction products evolve during the oxidation of chemically reduced organic aerosol. A kinetic model of multigenerational
H Schulz et al.
The European respiratory journal, 8(4), 566-573 (1995-04-01)
The contribution of aerosol techniques, the estimation of aerosol bolus dispersion and effective airway dimensions, to the clinical diagnosis of paediatric asthma was studied. In 47 children, aged 11 +/- 2 yrs, with mild asthma (forced expiratory volume in one
F J Wilson et al.
Journal of applied physiology (Bethesda, Md. : 1985), 58(1), 223-229 (1985-01-01)
Theoretical models of particle deposition in the respiratory tract predict high fractional deposition for particles of less than 0.1 micron, but there are few confirming experimental data for those predictions. We have measured the deposition fraction of a nonhygroscopic aerosol
P J Anderson et al.
The American review of respiratory disease, 144(3 Pt 1), 649-654 (1991-09-01)
How variable is the deposition of inhaled methacholine (MCH) in the respiratory tract during a challenge test? Does this variability contribute to the variability of airway responsiveness? To examine these questions we estimated the deposition of polydisperse MCH droplets by
C S Kim et al.
Journal of applied physiology (Bethesda, Md. : 1985), 81(5), 2203-2213 (1996-11-01)
Detailed regional deposition of inhaled particles was investigated in young adults (n = 11) by use of a serial bolus aerosol delivery technique. A small bolus (45 ml half-width) of monodisperse aerosols [1-, 3-, and 5-microns particle diameter (Dp)] was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service