Skip to Content
Merck
All Photos(1)

Key Documents

S2897

SAFC

EX-CELL® Hybri-Max Medium, Protein-Free

with L-glutamine, with MOPS buffer, with sodium bicarbonate, sterile-filtered, suitable for hybridoma

Synonym(s):

Hybridoma Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207

description

for research or for further manufacturing use
liquid

sterility

sterile-filtered

technique(s)

cell culture | hybridoma: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Formula variant

Does not require serum or other supplementation.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service