Skip to Content
Merck

Skip To

S1939

Anti-Na+/Ca2+/K+ Exchanger 2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NCKX2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 μL
Check Cart for Availability
€ 645,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
1

€ 645,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

rat

technique(s)

indirect immunofluorescence: suitable, western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

synthetic peptide corresponding to residues 90-102 of rat NCKX2.

Physical form

Solution in phosphate buffered saline containing 1.0 mg/ml bovine serum albumin and 0.05% sodium azide.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V9014SAB2103451SAB5200108
description

affinity isolated antibody, buffered aqueous solution

description

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

description

affinity isolated antibody

description

1 mg/mL, affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

Gene Information

rat ... Slc24a2(84550)

Gene Information

human ... SLC18A2(6571)
mouse ... Slc18a2(214084)
rat ... Slc18a2(25549)

Gene Information

human ... SLC23A2(9962)

Gene Information

rat ... Slc14a2(54302)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters