S1500
Sarcosine hydrochloride
Synonym(s):
N-Methylaminoacetic acid, N-Methylglycine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3NHCH2CO2H·HCl
CAS Number:
Molecular Weight:
125.55
EC Number:
MDL number:
UNSPSC Code:
12352200
mp
173-175 °C (lit.)
SMILES string
Cl.CNCC(O)=O
InChI
1S/C3H7NO2.ClH/c1-4-2-3(5)6;/h4H,2H2,1H3,(H,5,6);1H
InChI key
WVKIFIROCHIWAY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jonathan E McDunn et al.
The Prostate, 73(14), 1547-1560 (2013-07-05)
Current diagnostic techniques have increased the detection of prostate cancer; however, these tools inadequately stratify patients to minimize mortality. Recent studies have identified a biochemical signature of prostate cancer metastasis, including increased sarcosine abundance. This study examined the association of
Ondrej Zitka et al.
Electrophoresis, 34(18), 2639-2647 (2013-06-19)
Carcinoma of prostate (CaP) is the second most frequent malignant tumor occurring in men in Europe. Currently there is discussion on a wide range of potential CaP markers.One of them—nonprotein amino acid sarcosine, also known as N-methylglycine was chosen as
Simone J P M Eussen et al.
The British journal of nutrition, 110(2), 363-374 (2012-12-12)
Different lifestyle patterns across Europe may influence plasma concentrations of B-vitamins and one-carbon metabolites and their relation to chronic disease. Comparison of published data on one-carbon metabolites in Western European regions is difficult due to differences in sampling procedures and
Giuseppe Lucarelli et al.
The Journal of urology, 190(6), 2271-2277 (2013-05-15)
SPON2 belongs to the F-spondin family of secreted extracellular matrix proteins. It is deregulated in some tumors, including prostate cancer. In this prospective study we assessed the role of serum SPON2 as a biomarker for prostate cancer diagnosis as well
King Hang Aaron Lau et al.
Langmuir : the ACS journal of surfaces and colloids, 28(46), 16099-16107 (2012-10-30)
Poly(N-substituted glycine) "peptoids" are a class of peptidomimetic molecules receiving significant interest as engineered biomolecules. Sarcosine (i.e., poly(N-methyl glycine)) has the simplest side chain chemical structure of this family. In this Article, we demonstrate that surface-grafted polysarcosine (PSAR) brushes exhibit
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service