S150
Spermine–Nitric oxide complex
source of nitric oxide
Synonym(s):
N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl]-1,3-propanediamine, SPER/NO, Spermine NONOate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
powder
storage condition
desiccated
color
white
solubility
methanol: ≥1 mg/mL
H2O: ≥2 mg/mL
εmax
8500 at 252 nm
shipped in
dry ice
storage temp.
−20°C
SMILES string
NCCCN(N(N=O)O)CCCCNCCCN
InChI
1S/C10H26N6O2/c11-5-3-8-13-7-1-2-9-15(10-4-6-12)16(18)14-17/h13,18H,1-12H2
InChI key
QPIOUFBAROSIAN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Used to produce a controlled release of nitric oxide in solution.
Other Notes
air sensitive
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cancer letters, 273(1), 148-154 (2008-09-12)
Here we demonstrate a synergistic and tumor selective cytotoxic effect by combined treatment with naphthoxylosides, polyamine synthesis inhibitor, and polyamine based nitric oxide (NO) donor, using in vitro human tumor models. We have earlier reported that heparan sulfate priming naphthoxyloside
Scientific reports, 7(1), 15083-15083 (2017-11-10)
The PhoPQ two-component regulatory system coordinates the response of Salmonella enterica serovar Typhimurium to diverse environmental challenges encountered during infection of hosts, including changes in Mg
Metabolism: clinical and experimental, 62(1), 79-89 (2012-08-15)
The amino acid Arginine (Arg) is the main biological precursor of nitric oxide (NO) and has been described to improve insulin sensitivity in diabetes and obesity. We investigated the molecular mechanisms involved in the long-term effects of Arg on glucose
Glia, 58(9), 1133-1144 (2010-05-15)
To understand the pathomechanisms of spinal cord injuries will be a prerequisite to develop efficient therapies. By investigating acute lesions of spinal cord white matter in anesthetized mice with fluorescently labeled microglia and axons using in vivo two-photon laser-scanning microscopy
Diabetologia, 53(6), 1142-1150 (2010-03-30)
We investigated the direct effect of a nitric oxide donor (spermine NONOate) on glucose transport in isolated human skeletal muscle and L6 skeletal muscle cells. We hypothesised that pharmacological treatment of human skeletal muscle with N-(2-aminoethyl)-N-(2-hydroxy-2-nitrosohydrazino)-1,2-ethylenediamine (spermine NONOate) would increase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service