Skip to Content
Merck
All Photos(1)

Key Documents

S0689

Sigma-Aldrich

Anti-Serotonin N-Acetyltransferase (C-Terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€339.00

€339.00


Check Cart for Availability


Select a Size

Change View
50 μG
€339.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€339.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

mouse, rat

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Aanat(11298)
rat ... Aanat(25120)

Immunogen

synthetic phosphopeptide corresponding to carboxy-terminal amino acid residues 198-208 of rat arylalkylamine N-acetyltransferase conjugated to KLH.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml bovine serum albumin and 0.05% sodium azide

Preparation Note

Non-phospho-specific antibody was isolated on non-phospho-peptide coupled resin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Marcia Muxel et al.
PloS one, 7(12), e52010-e52010 (2013-01-04)
We demonstrate that during inflammatory responses the nuclear factor kappa B (NF-κB) induces the synthesis of melatonin by macrophages and that macrophage-synthesized melatonin modulates the function of these professional phagocytes in an autocrine manner. Expression of a DsRed2 fluorescent reporter
Claudia Emanuele Carvalho-Sousa et al.
Journal of pineal research, 68(3), e12636-e12636 (2020-02-12)
Environmental pollution in the form of particulate matter <2.5 μm (PM2.5 ) is a major risk factor for diseases such as lung cancer, chronic respiratory infections, and major cardiovascular diseases. Our goal was to show that PM2.5 eliciting a proinflammatory response

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service