Skip to Content
Merck
All Photos(1)

Documents

S0564

Sigma-Aldrich

Anti-Serotonin N-Acetyltransferase (N-Terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AANAT(15)
mouse ... Aanat(11298)
rat ... Aanat(25120)

Immunogen

synthetic phosphopeptide corresponding to amino-terminal amino acid residues 24-33 of mouse arylalkylamine N-acetyltransferase conjugated to KLH.

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml bovine serum albumin and 0.05% sodium azide

Preparation Note

The non-phospho-specific antibody was isolated on non-phospho-peptide-coupled resin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco A Pires-Lapa et al.
International journal of molecular sciences, 19(8) (2018-07-28)
Melatonin (5-methoxy-N-acetylserotonin), the pineal hormone, is also synthesized by immune-competent cells. The pineal hormone signals darkness, while melatonin synthesized on demand by activated macrophages at any hour of the day acts locally, favoring regulatory/tolerant phenotypes. Activation of β-adrenoceptors in pinealocytes
Marlina O Córdoba-Moreno et al.
Scientific reports, 10(1), 4799-4799 (2020-03-18)
Daily oscillation of the immune system follows the central biological clock outputs control such as melatonin produced by the pineal gland. Despite the literature showing that melatonin is also synthesized by macrophages and T lymphocytes, no information is available regarding
Claudia Emanuele Carvalho-Sousa et al.
Journal of pineal research, 68(3), e12636-e12636 (2020-02-12)
Environmental pollution in the form of particulate matter <2.5 μm (PM2.5 ) is a major risk factor for diseases such as lung cancer, chronic respiratory infections, and major cardiovascular diseases. Our goal was to show that PM2.5 eliciting a proinflammatory response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service