Skip to Content
Merck
All Photos(1)

Key Documents

S0274

Sigma-Aldrich

[D-Arg1, D-Trp7,9, Leu11]-Substance P acetate salt

≥97% (HPLC)

Synonym(s):

Spantide I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H108N20O13 · xC2H4O2
CAS Number:
Molecular Weight:
1497.79 (free base basis)
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]6CCCN6C(=O)[C@H](CCCCN)NC(=O)[C@@H]7CCCN7C(=O)[C@H](N)CCCNC(N)=N)C(N)=O

Gene Information

human ... TACR1(6869)
mouse ... TACR1(21336)
rat ... TACR1(24807)

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Trp-Phe-Trp-Leu-Leu-NH2

Biochem/physiol Actions

Potent NK-1 tachykinin receptor antagonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon A McClellan et al.
Investigative ophthalmology & visual science, 49(4), 1502-1511 (2008-04-04)
Studies have shown that blocking substance P (SP) binding to neurokinin 1 receptor with spantide I prevents Pseudomonas aeruginosa-induced corneal perforation in susceptible C57BL/6 mice. This study tested the effect of SP injection on the resistance response (cornea heals) of
Zimei Zhou et al.
Investigative ophthalmology & visual science, 49(10), 4458-4467 (2008-06-21)
Apoptosis was examined after Pseudomonas aeruginosa corneal infection in C57BL/6 (B6, susceptible) and BALB/c (resistant) mice. TUNEL staining, real-time RT-PCR, polymorphonuclear neutrophils (PMNs) and macrophage (Mphi) depletion, and immunostaining were used. Intense TUNEL staining was seen in BALB/c versus B6
A C Broiz et al.
Neuroscience, 201, 134-145 (2011-11-30)
The dorsal periaqueductal gray (dPAG) is the main output structure for the defensive response to proximal aversive stimulation. Panic-like responses, such as freezing and escape behaviors, often result when this structure is electrically stimulated. Freezing also ensues after termination of
Jeffrey Tessier et al.
Infection and immunity, 75(4), 1895-1903 (2007-01-31)
Bacillus anthracis edema toxin (ET), composed of protective antigen and an adenylate cyclase edema factor (EF), elicits edema in host tissues, but the target cells and events leading from EF-mediated cyclic-AMP production to edema are unknown. We evaluated the direct
Radomir Radomirov et al.
Archives italiennes de biologie, 148(4), 377-388 (2011-02-11)
The evacuative motor responses of the anal canal and recto-anal reflexes during defecation were studied in an isolated rat recto-anal model preparation using (i) partitioned organ bath, (ii) electrical stimulation, (iii) balloon distension and (iv) morphological techniques. Electrical field stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service