Skip to Content
Merck
All Photos(4)

Documents

RNB500

Sigma-Aldrich

GenElute Plasma/Serum RNA Purification Mini Kit

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute Plasma/Serum RNA Purification Mini Kit provides a fast, reliable, reproducible and simple procedure for isolating circulating RNA and exosomal RNA from small plasma/serum inputs ranging from 10 μL to 25 μL, with various kit formats addressing different plasma/serum input volumes. Purification is based on spin column chromatography that uses Sigma′s proprietary resin separation matrix. The kit is designed to isolate all sizes of circulating RNA, including microRNA, as well as all sizes of exosomal RNA.

Sigma′s GenElute Plasma/Serum RNA Purification Mini Kit provides a clear advantage over other available kits in that they do not require phenol/chloroform or any protease treatments. RNA can be isolated from either fresh or frozen samples using these kits. Moreover, the kits allow the user to elute into a flexible elution volume ranging from 10 μL to 100 μL. Typical yields of free-circulating and exosomal RNA vary depending on the input sample, as the amount of RNA present in plasma and serum will depend upon the health status of the individual.

  • Isolate all sizes of circulating and exosomal RNA, including microRNA
  • Versatile plasma/serum input ranging from 50 μL to 200 μL
  • No phenol extractions
  • No carrier RNA
  • Concentrate circulating RNA and Exosomal RNA into a flexible elution volume ranging from 10 μL to 25 μL
  • Purify high-quality RNA in 15-20 minutes

Application

  • Reverse transcription qPCR
  • Reverse transcription PCR
  • Northern blotting
  • RNase protection and primer extension
  • Expression array assays.
  • miRNA array
  • RNA-Seq
  • Nanostring
  • Fluidigm
  • Droplet/Digital PCR

Biochem/physiol Actions

GenElute Plasma/Serum RNA Purification Mini Kit has been used for extraction of total serum RNA from human hepatocellular carcinoma (HCC) and colorectal cancer (CRC) samples.

Unit Definition

Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MicroRNA-144 mediates chronic inflammation and tumorigenesis in colorectal cancer progression via regulating C-X-C motif chemokine ligand 11
Han B, et al.
Experimental and Therapeutic Medicine, 16(3), 1935-1943 (2018)
Wenwen Yang et al.
Medicine, 99(47), e23255-e23255 (2020-11-22)
The study aims to characterize the circular RNA (circRNA) expression profile that is functionally related with the invasiveness of papillary thyroid microcarcinoma (PTMC).A total of 13 pairs of female patients with non-invasive PTMC or lymph node metastasis PTMC (PTMC (L))
Yuhua Gong et al.
Cancer cell international, 18, 116-116 (2018-08-21)
Accumulating data indicated that circRNA plays important roles in regulating many biological processes of the tumor, the present study is designated for exploring roles of the circ-ZEB1.33-miR-200a-3p-CDK6 regulating axis in human hepatocellular carcinoma (HCC). The regulation axis as predicted by
Circ-ZEB1. 33 promotes the proliferation of human HCC by sponging miR-200a-3p and upregulating CDK6
Gong Y, et al.
Cancer Cell International, 18(1), 116-116 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service