Key Documents
RF45645P-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-64-5-PCR, non aerosol tight, to hold, 64 x 0.2 mL PCR tubes, lid
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
feature
lid
to hold 64 x 0.2 mL PCR tubes
non aerosol tight
strip rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022654209 (NA)
Eppendorf® 5427714006 (EU)
parameter
13,543 × g max. RCF
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)