Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Check Cart for Availability | € 848,00 |
About This Item
sample type cell culture supernatant(s)
sample type plasma
€ 848,00
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.410-100 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Fgfbp1(64535)
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for cell culture supernatants, plasma, and serum samples | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants |
| species reactivity rat | species reactivity rat | species reactivity rat | species reactivity rat |
| Gene Information rat ... Fgfbp1(64535) | Gene Information rat ... Fgf2(54250) | Gene Information rat ... Flt1(54251) | Gene Information rat ... Il9(116558) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type serum | input sample type cell culture supernatant(s) | input sample type cell culture supernatant(s) | input sample type plasma |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




