Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | € 848,00 |
About This Item
sample type plasma
sample type serum
€ 848,00
species reactivity
porcine
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
Porcine ... IGFBP5(397182)
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for serum, plasma, cell culture supernatant and urine | description - | description for serum, plasma, cell culture supernatant and urine |
| species reactivity porcine | species reactivity human | species reactivity rat | species reactivity human |
| Gene Information Porcine ... IGFBP5(397182) | Gene Information human ... IGFBP5(3488) | Gene Information rat ... Igfbp5(25285) | Gene Information human ... IGFBP1(3484) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type cell culture supernatant(s) | input sample type cell culture supernatant(s) | input sample type cell culture supernatant(s) | input sample type plasma |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable | technique(s) capture ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




