Skip to Content
Merck
All Photos(1)

Key Documents

RAB1678

Sigma-Aldrich

Human SCARA5 ELISA

for serum, plasma and cell culture supernatants

Synonym(s):

Scavenger receptor class A member 5 , Scavenger receptor hlg

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€831.00

€831.00


Check Cart for Availability


Select a Size

Change View
1 KIT
€831.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€831.00


Check Cart for Availability

species reactivity

human

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SCARA5(286133)

General description

Scavenger receptor class A member 5 (SCARA5) belongs to the scavenger receptor family. It is an L-ferritin receptor that mediates non-transferrin iron delivery in kidney organogenesis.[1] SCARA5 is generally expressed in the kidney, bladder, ovary, adrenal gland, testis, skin, and trachea. It serves as a tumor suppressor in several human cancers.[2]

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damla Ulker et al.
Gene, 673, 102-106 (2018-06-17)
Breast cancer is the most common malignant tumor in women worldwide. Breast tumors mostly exhibit aberrant gene expression and DNA hypermethylation patterns that predispose the disease. Understanding the genetic and epigenetic factors that contribute to breast cancer development is important
Chen Zheng et al.
Cell biochemistry and function, 38(2), 158-166 (2020-01-29)
Thyroid cancer (TC) has become one of most common endocrine malignancies in recent decades. Due to gene background polymorphism, it's outcome goes quite differently in each patient. For exploring the mechanism, we performed whole transcriptome sequencing of paired papillary thyroid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service