Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | € 848,00 |
About This Item
sample type plasma
sample type serum
€ 848,00
species reactivity
human
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CD5(921)
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description clone CRIS1, purified immunoglobulin, buffered aqueous solution | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants |
| Gene Information human ... CD5(921) | Gene Information human ... CD5(921) | Gene Information human ... CD5L(922) | Gene Information human ... CD6(923) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| assay range inter-assay cv: <12% | assay range - | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type cell culture supernatant(s) | input - | input sample type cell culture supernatant(s) | input sample type serum |
| technique(s) ELISA: suitable | technique(s) flow cytometry: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




