Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery) | € 637,00 |
About This Item
sample type plasma
sample type cell culture supernatant(s)
€ 637,00
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum sample(s)
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.8 ng/ml
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Hgf(24446)
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information rat ... Hgf(24446) | Gene Information mouse ... Hgf(15234) | Gene Information mouse ... Hgf(15234) | Gene Information human ... HGF(3082) |
| species reactivity rat | species reactivity mouse | species reactivity mouse | species reactivity human |
| input sample type serum sample(s) | input sample type cell lysate | input sample type plasma | input sample type tissue lysate |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




