Skip to Content
Merck
All Photos(1)

Key Documents

RAB1127

Sigma-Aldrich

Mouse PAI-1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€624.00

€624.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
€624.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€624.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

mouse, human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/ml

shipped in

wet ice

storage temp.

−20°C

General description

The PAI-1 (plasminogen activator inhibitor) gene is localized to human chromosome 7q22.1.[1] Active form of PAI-1 is expressed in the blood and tissues. Platelets harbor both the active and latent forms of PAI-1.[2]
This ELISA antibody pair detects mouse and human PAI-1.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

PAI-1 (plasminogen activator inhibitor) produced by platelets takes part in stabilizing blood clotting mechanism. PAI-1 prevents plasminogen activation during fibrinolysis. This might lead to the risk of cardiovascular disease. Elevated levels of PAI-1 has been observed in type 2 diabetes mellitus and serves the reason for reduced insulin sensitivity.[3] PAI-1 stimulates the process of cellular senescence. PAI-1 regulation occurs through p53 dependent TGF-β (transforming growth factor-β) signalling.[4] Studies also also predict that stress and depression might influence PAI-1 expression.[5]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperglycaemia-induced reciprocal changes in miR-30c and PAI-1 expression in platelets.
Luo M, et al.
Scientific Reports, 6, 36687-36687 (2016)
Plasminogen activator inhibitor-1 in depression: results from animal and clinical studies.
Jiang H, et al.
Scientific Reports, 6, 30464-30464 (2016)
Hemostasis and Thrombosis: Basic Principles and Clinical Practice, 366-366 (2006)
TGF-? induces p53/Smads complex formation in the PAI-1 promoter to activate transcription.
Kawarada Y, et al.
Scientific Reports, 6, 35483-35483 (2016)
CpG methylation of the PAI-1 gene 5?-flanking region is inversely correlated with PAI-1 mRNA levels in human cell lines.
Gao S, et al.
Thrombosis and Haemostasis, 93(03), 651-660 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service