Skip to Content
Merck

Skip To

RAB1126

Mouse Osteoactivin ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 576,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Gpnmb(93695)
Input:
sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

€ 576,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Gpnmb(93695)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1822RAB1011RAB1017
description

for cell culture supernatants, plasma, and serum samples

description

for serum, plasma and cell culture supernatants

description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

Gene Information

mouse ... Gpnmb(93695)

Gene Information

-

Gene Information

-

Gene Information

-

species reactivity

mouse

species reactivity

rhesus macaque

species reactivity

bovine

species reactivity

canine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.2 ng/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/ml

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

input

-

input

-

input

-

detection method

colorimetric

detection method

-

detection method

-

detection method

-

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters