Skip to Content
Merck
All Photos(1)

Key Documents

RAB1124

Sigma-Aldrich

Mouse Gremlin-1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.8 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Grem1(23892)

General description

Gremlin-1 (GREM1) is part of the secreted, cysteine knot DAN family of bone morphogenetic protein antagonists. It has a molecular weight of 24–26kDa.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Gremlin-1 (GREM1) has a role in embryogenesis. It also has a role in cell functions during development. GREM1 acts as an antagonist of bone morphogenetic proteins (BMP). The protein influences chondrogenesis, outgrowth and apoptosis in limb bud development and has a role in kidney formation. It inhibits the activity of BMP2 and BMP4. GREM1 is involved in angiogenesis, bone formation, bone mass and pulmonary hypertension. Knockout of this gene in mice leads to developmental abnormalities.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Treatment with anti-gremlin 1 antibody ameliorates chronic hypoxia/SU5416-induced pulmonary arterial hypertension in mice.
Ciuclan L, et al.
The American Journal of Pathology, 183(5), 1461-1473 (2013)
Liukun Meng et al.
Journal of the American Heart Association, 9(15), e016586-e016586 (2020-08-05)
Background Heterozygous mutation in BMP (bone morphogenetic protein) receptor 2 is rare, but BMP cascade suppression is common in congenital heart disease-associated pulmonary arterial hypertension (CHD-PAH); however, the underling mechanism of BMP cascade suppression independent of BMP receptor 2 mutation
Gremlin1 plays a key role in kidney development and renal fibrosis.
Church RH, et al.
American Journal of Physiology: Renal Physiology, 312(6), F1141-F1157 (2017)
Allelic Depletion of grem1 Attenuates Diabetic Kidney Disease
Roxburgh SA, et al.
Diabetes, 58(7), 1641-1650 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service