Skip to Content
Merck

Skip To

RAB1102

Rhesus Macaque IL-6R ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 539,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
human, rhesus macaque
Storage temp.:
−20°C
Shipped in:
wet ice

€ 539,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human, rhesus macaque

technique(s)

ELISA: suitable

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/ml

shipped in

wet ice

storage temp.

−20°C

General description

This ELISA antibody pair detects Rhesus Macaque and human IL-6R. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1101RAB1103RAB1809
description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

description

for serum, plasma and cell culture supernatants

species reactivity

human, rhesus macaque

species reactivity

rhesus macaque

species reactivity

rhesus macaque

species reactivity

rhesus macaque

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.2 ng/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.4 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.045 ng/mL
standard curve range: 0.041-10 ng/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters