Skip to Content
Merck
All Photos(1)

Key Documents

RAB1071

Sigma-Aldrich

Human MDK ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.12 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MDK(4192)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Zhang et al.
Journal of clinical laboratory analysis, 32(8), e22576-e22576 (2018-05-26)
Midkine (MK) level has been shown to be elevated in serum of patients with nonsmall cell lung cancer (NSCLC). However, the diagnostic value of MK in pleural effusion in NSCLC has not been well validated and established. Samples of NSCLC-associated
Carolina Cubillos-Zapata et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(12), 16179-16190 (2020-10-16)
Midkine (MDK) might mediate the proangiogenic effect of intermittent hypoxia (IH) in patients with obstructive sleep apnea (OSA) and cutaneous melanoma (CM). We compare circulating MDK in CM patients with and without OSA, and their relationship with tumor aggressiveness, while
Jeffrey Wang et al.
Scientific reports, 10(1), 14499-14499 (2020-09-04)
Midkine (MDK), a heparin-binding growth factor cytokine, is involved in the pathogenesis of kidney diseases by augmenting leukocyte trafficking and activation. Animal models and small case control studies have implicated MDK as a pathological biomarker in chronic kidney diseases (CKD)
Youjia Han et al.
Cancer letters, 445, 24-33 (2019-01-07)
Medulloblastoma is the most prevalent central nervous system tumor in children. Targeted treatment approaches for patients with high-risk medulloblastoma are needed as current treatment regimens are not curative in many cases and cause significant therapy-related morbidity. Medulloblastoma harboring MYC amplification
Ina Lackner et al.
Frontiers in immunology, 10, 1920-1920 (2019-09-26)
Background and Purpose: Post-traumatic cardiac dysfunction often occurs in multiply injured patients (ISS ≥ 16). Next to direct cardiac injury, post-traumatic cardiac dysfunction is mostly induced by the release of inflammatory biomarkers. One of these is the heparin-binding factor Midkine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service