Sign In to View Organizational & Contract Pricing.
Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*
Sign in to check eligibility. Shop Products
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | € 545,00 |
About This Item
NACRES:
NA.32
UNSPSC Code:
41116158
€ 545,00
Save Up To 15 %
Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
species reactivity
human
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... EGFR(1956)
General description
The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-EGFR.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Kit Components Also Available Separately
Product No.
Description
SDS & Pricing
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
