Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | € 858,00 |
About This Item
€ 858,00
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Il17f(257630)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information mouse ... Il17f(257630) | Gene Information mouse ... Il7r(16197) | Gene Information mouse ... Il17rb(50905) | Gene Information mouse ... Il2Ra(16184) |
| species reactivity mouse | species reactivity mouse | species reactivity mouse | species reactivity mouse |
| assay range inter-assay cv: <10% | assay range inter-assay cv: <10% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| packaging kit of 96 wells (12 strips x 8 wells) | packaging kit of 96 wells (12 strips x 8 wells) | packaging kit of 96 wells (12 strips x 8 wells) | packaging kit of 96 wells (12 strips x 8 wells) |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




