RAB0780
Mouse Tyro3 / Tyrosine-protein Kinase Receptor TYRO3 ELISA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.4 ng/mL
standard curve range: 0.416-100 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Tyro3(22174)
Related Categories
General description
The antibody pair provided in this kit recognizes mouse Tyro3.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Tyrosine-protein kinase receptor (TYRO3) is a transmembrane tyrosine kinase receptor. It plays a significant role in the brain development during embryogenesis. It plays an important role in apoptotic cell clearance. In hepatocellular carcinoma, it is associated with tumor growth, aggressiveness and liver dysfunction. Mutations in the TYRO3 gene intronic regions are linked with asthma. TYRO3 is also a suppressor of type 2 immunity. As part of the TAM (TYRO3, AXL and MER receptors) family, it is activated by PROS1 (protein S) and GAS6 (growth arrest-specific gene 6). Changes in the expression of TYRO3 are linked with the proliferation and survival of cancer cells. Absence of TYRO3 in mice results in neurological disorders.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Gas6/TAM System and Multiple Sclerosis.
Bellan M, et al.
International Journal of Molecular Sciences, 17 (2016)
Increased soluble phagocytic receptors sMer, sTyro3 and sAxl and reduced phagocytosis in juvenile-onset systemic lupus erythematosus.
Ballantine L, et al.
Pediatric Rheumatology Online Journal (2015)
The TAM family receptor tyrosine kinase TYRO3 is a negative regulator of type 2 immunity.
Chan PY
Science, 352, 99-103 (2016)
Overexpression of Tyro3 and its implications on hepatocellular carcinoma progression.
Duan Y
International Journal of Oncology, 48, 358-366 (2016)
Receptor tyrosine kinases, TYRO3, AXL, and MER, demonstrate distinct patterns and complex regulation of ligand-induced activation.
Tsou WI
The Journal of Biological Chemistry, 289, 25750-25763 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service