Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | € 637,00 |
About This Item
sample type plasma
sample type cell culture supernatant(s)
€ 637,00
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 4.1-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Tgfbi(21810)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description targeting mouse Tgfbi | description for serum, plasma, cell culture supernatant and urine | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| Gene Information mouse ... Tgfbi(21810) | Gene Information mouse ... Tgfbi(21810) | Gene Information human ... TGFBI(7045) | Gene Information human ... TGFBI(7045) |
| species reactivity mouse | species reactivity - | species reactivity human | species reactivity human |
| input sample type serum | input - | input sample type urine | input - |
| assay range inter-assay cv: <12% | assay range - | assay range inter-assay cv: <12% | assay range - |
| detection method colorimetric | detection method - | detection method colorimetric | detection method - |
| technique(s) ELISA: suitable | technique(s) - | technique(s) capture ELISA: suitable | technique(s) immunohistochemistry: 1:500- 1:1000 |
Kit Components Also Available Separately
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




