Skip to Content
Merck

Skip To

RAB0660

Human FSH ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 539,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 539,00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

The antibody pair provided in this kit recognizes human FSH.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Follicle stimulating hormone (FSH) belongs to glycoprotein hormone family. It is secreted by the gonadotropic cells of pituitary gland. FSH specifically binds to FSH receptor (FSHR) for signalling. It also plays an important role in oogenesis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0737RAB1935RAB0677
description

-

description

-

description

for serum, plasma and cell culture supernatants

description

-

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 ng/mL
standard curve range: 0.328-80 ng/mL

assay range

inter-assay cv: 12%
intra-assay cv: 10%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters