Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | € 660,00 |
About This Item
sample type plasma
sample type cell culture supernatant(s)
€ 660,00
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.025 ng/mL
standard curve range: 0.027-20 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... APOA2(336)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| Gene Information human ... APOA2(336) | Gene Information human ... APOB(338) | Gene Information human ... APOC3(345) | Gene Information human ... APOC2(344) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type serum | input sample type cell culture supernatant(s) | input sample type plasma | input sample type serum |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




