Skip to Content
Merck

Skip To

RAB0533

Human CLU / Clusterin ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

Apo J, Apolipoprotein J, Clusterin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 539,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... CLU(1191)
Input:
sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 539,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 15 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLU(1191)

General description

This ELISA antibody pair detect human Clusterin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB3500199RAB0775RAB1793
description

for serum, plasma, cell culture supernatants and urine

description

affinity isolated antibody, buffered aqueous solution

description

-

description

for serum, plasma and cell culture supernatants

species reactivity

human

species reactivity

human, mouse

species reactivity

mouse

species reactivity

rhesus macaque

Gene Information

human ... CLU(1191)

Gene Information

human ... CLU(1191)

Gene Information

mouse ... Clu(12759)

Gene Information

rhesus macaque ... CLU(677866)

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 15 pg/mL

assay range

-

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.1 ng/mL
standard curve range: 0.098-24 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

input

-

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

technique(s)

ELISA: suitable

technique(s)

immunohistochemistry: suitable, indirect ELISA: suitable, immunofluorescence: suitable, western blot: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

-

detection method

colorimetric

detection method

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer E (Item E2)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters