Skip to Content
Merck

Skip To

RAB0491

Human CD30 ELISA Kit

for serum, plasma and cell culture supernatant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 479,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... TNFRSF8(943)
Input:
sample type serum
sample type cell culture supernatant(s)
sample type plasma
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 479,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.48-5000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF8(943)

General description

The Human CD30 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CD30 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human CD30

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB56000325SAB4700151RAB0492
description

for serum, plasma and cell culture supernatant

description

clone RM425, affinity purified immunoglobulin

description

clone MEM-268, purified immunoglobulin, buffered aqueous solution

description

for serum, plasma and cell culture supernatant

Gene Information

human ... TNFRSF8(943)

Gene Information

human ... TNFRSF8(943)

Gene Information

human ... TNFRSF8(943)

Gene Information

mouse ... Tnfrsf8(21941)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.48-5000 pg/mL

assay range

-

assay range

-

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 12.29-3000 pg/mL

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

-

input

-

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

immunohistochemistry: 1:100-1:250 , western blot: 1:1000-1:2500

technique(s)

flow cytometry: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

detection method

colorimetric

detection method

-

detection method

-

detection method

colorimetric


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters