Skip to Content
Merck
All Photos(2)

Documents

RAB0359

Sigma-Aldrich

Human MICB ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 75 pg/mL
standard curve range: 0.0686-50 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MICB(4277)

General description

The Human MICB (MHC class I Chain-related gene B) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human MICB in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MICB

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel H Libraty et al.
PloS one, 9(5), e98509-e98509 (2014-05-30)
Dengue is the most prevalent arthropod-borne viral illness in humans. A MHC class I polypeptide-related sequence B (MICB) single nucleotide polymorphism (SNP) was previously associated with symptomatic dengue compared to non-dengue causes of acute febrile illnesses in infants. We measured
Susanne H Baumeister et al.
Cancer immunology research, 7(1), 100-112 (2018-11-07)
NKG2D ligands are widely expressed in solid and hematologic malignancies but absent or poorly expressed on healthy tissues. We conducted a phase I dose-escalation study to evaluate the safety and feasibility of a single infusion of NKG2D-chimeric antigen receptor (CAR)
Jing Bian et al.
Life sciences, 246, 117396-117396 (2020-02-09)
Hepatocellular carcinoma (HCC) is a leading cause of cancer mortality worldwide. Decrease in NKG2D ligand levels and exhaustion of NK cells in HCC patients are major causes of immune escape, high recurrence, poor prognosis, and low overall survival. Enhancing the
Kelly C G ManfrereC et al.
Oncotarget, 8(54), 92183-92194 (2017-12-02)
Sézary syndrome (SS), an aggressive and leukemic form of cutaneous T-cell lymphoma, usually results in shortened survival. Improving innate immunity in SS by targeting natural killer (NK) cells with Toll-like receptor (TLR) agonists could be an interesting modulatory strategy. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service