Skip to Content
Merck

Skip To

RAB0146

Human Dkk-4 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 545,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... DKK4(27121)
Input:
sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 545,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 13.72-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DKK4(27121)

General description

The Human Dkk-4 (Dickkopf homolog-4) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Dkk-4 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human DKK4

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0144RAB0237RAB0298
description

for serum, plasma, cell culture supernatants and urine

description

for serum, plasma and cell culture supernatant

description

for serum, plasma, cell culture supernatant and urine

description

for serum, plasma, cell culture supernatant and urine

Gene Information

human ... DKK4(27121)

Gene Information

mouse ... Dkk1(13380)

Gene Information

human ... IGFBP4(3487)

Gene Information

human ... IL4(3565)

species reactivity

human

species reactivity

mouse

species reactivity

human

species reactivity

human

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

input

sample type serum
sample type urine
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 13.72-10000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 24.58-6000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 250 pg/mL
standard curve range: 0.41-100 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 3.13-200 pg/mL

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer E (Item E2)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters