Skip to Content
Merck
All Photos(2)

Documents

RAB0072

Sigma-Aldrich

Human CCL28 / VIC ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 27.4-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL28(56477)

General description

The Human CCL28 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CCL28 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human CCL28

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manon Nayrac et al.
Mucosal immunology, 14(1), 219-228 (2020-04-30)
Gut CD4+ T cells are incompletely restored in most HIV-1-infected individuals on antiretroviral therapy, notably Th17 cells, a key subset in mucosal homeostasis. By contrast, gut Th22 cells are usually restored at normal frequencies. Th22 cells display a CCR6+CCR10+ phenotype
Yingting Wu et al.
Molecular medicine reports, 25(2) (2021-12-17)
Endometriosis (EM), the presence of functional endometrial glands and stroma outside the uterine cavity, is a common gynecological disorder. At present, the pathogenesis of EM has not been fully elucidated, so there is still a lack of effective therapy. The
Hongzhe Li et al.
Haematologica, 105(5), 1206-1215 (2019-08-03)
Human bone marrow stromal cells (BMSC) are key elements of the hematopoietic environment and they play a central role in bone and bone marrow physiology. However, how key stromal cell functions are regulated is largely unknown. We analyzed the role
Amanda M Burkhardt et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 39(4), 214-223 (2019-03-12)
CCL28 is a mucosal chemokine that has been involved in various responses, including IgA production. We have analyzed its production in human tissues using a comprehensive microarray database. Its highest expression is in the salivary gland, indicating that it is
Lu Ji et al.
Cancer research, 80(10), 2004-2016 (2020-03-12)
Dysregulation of Wnt/β-catenin signaling is frequently observed in human gastric cancer. Elucidation of the tumor immune microenvironment is essential for understanding tumorigenesis and for the development of immunotherapeutic strategies. However, it remains unclear how β-catenin signaling regulates the tumor immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service