Skip to Content
Merck
All Photos(1)

Documents

R9157

Sigma-Aldrich

Ro 4-1284

≥98% (HPLC)

Synonym(s):

2-Hydroxy-2-ethyl-3-isobutyl-9,10-dimethoxy-1,2,3,4,5,6,7-hexahydrobenzo[a]chinolizine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33NO3
CAS Number:
Molecular Weight:
347.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 16 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

CCC1(O)CC2N(CCc3cc(OC)c(OC)cc23)CC1CC(C)C

InChI

1S/C21H33NO3/c1-6-21(23)12-18-17-11-20(25-5)19(24-4)10-15(17)7-8-22(18)13-16(21)9-14(2)3/h10-11,14,16,18,23H,6-9,12-13H2,1-5H3

InChI key

TUNMGCULOKMBNJ-UHFFFAOYSA-N

Biochem/physiol Actions

Ro 4-1284 is a reversible VMAT2 inhibitor.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Leysen et al.
European journal of pharmacology, 206(1), 39-45 (1991-01-25)
[3H]Ketanserin was found to label (besides a low amount of 5-HT2 receptors) non-serotonergic binding sites on human platelet membranes. The latter binding was detected in the presence of excess of the 5-HT2 antagonist BW501, and was potently inhibited by tetrabenazine.
A M Dopico et al.
Journal of neurochemistry, 61(2), 481-489 (1993-08-01)
A single dose of x-irradiation was applied on the cephalic end of newborn rats, and the alterations in the noradrenergic afferents to the cerebellum were studied 180 days later. A net increase in the noradrenaline content of cerebellum was found
C R Berry et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 37(12), 2011-2016 (1996-12-01)
Para-[18F]fluorobenzylguanidine ([18F]PFBG) is a newly developed tracer for imaging myocardial sympathetic neuronal innervation. This study investigated the uptake and retention mechanisms of [18F]PFBG in perfused, isolated rat heart. Fluorine-18-PFBG was administered to working rat hearts within the perfusion medium at
E Missirlis et al.
Investigational new drugs, 8(2), 145-147 (1990-05-01)
Inhibition of angiogenesis offers an alternative approach to cancer chemotherapy, since solid tumor growth has an absolute dependency on angiogenesis. We have previously shown that 8,9-dihydroxy-7-methyl-benzo [b]quinolizinium bromide (GPA1734) is a basement membrane synthesis inhibitor, and that this compound acts
Innately hearing impaired Sprague-Dawley rats exhibit audiogenic seizure susceptibility following monoamine depletion.
C E Reigel et al.
Proceedings of the Western Pharmacology Society, 36, 267-271 (1993-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service