Skip to Content
Merck
All Photos(1)

Key Documents

R8900

Sigma-Aldrich

Ro 8-4304

solid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€151.00

€151.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 MG
€151.00

About This Item

Empirical Formula (Hill Notation):
C21H23FN2O3 · HCl
Molecular Weight:
406.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€151.00


Estimated to ship on25 May 2025


form

solid

solubility

H2O: ≤0.5 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

Cl.NC(=O)c1ccc(OCCC(O)N2CCC(=CC2)c3ccc(F)cc3)cc1

InChI

1S/C21H23FN2O3.ClH/c22-18-5-1-15(2-6-18)16-9-12-24(13-10-16)20(25)11-14-27-19-7-3-17(4-8-19)21(23)26;/h1-9,20,25H,10-14H2,(H2,23,26);1H

InChI key

NYFXHCMXYODFLP-UHFFFAOYSA-N

Gene Information

human ... GRIN2B(2904)

Biochem/physiol Actions

NR2B-selective, noncompetitive NMDA glutamate receptor antagonist

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Huang et al.
Molecular plant, 9(12), 1620-1633 (2016-10-21)
Nucleotide-binding leucine-rich repeat (NLR) proteins serve as immune receptors in both plants and animals. To identify components required for NLR-mediated immunity, we designed and carried out a chemical genetics screen to search for small molecules that can alter immune responses
Ghelardini, C., et al.
Drug Development Research, 40, 251-258 (1997)
Neuronal nicotinic acetylcholine receptors as novel drug targets.
G K Lloyd et al.
The Journal of pharmacology and experimental therapeutics, 292(2), 461-467 (2000-01-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service