Skip to Content
Merck
All Photos(1)

Documents

R8029

Sigma-Aldrich

Anti-RAD17 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-S.cerevisiae homolog of RAD24

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 70 kDa (also recognizes 75 kDa)

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect immunofluorescence: 5-10 μg/mL using HEK293-T cells fixed with methanol-acetone
microarray: suitable
western blot: 0.5-1.0 μg/mL using HeLa cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD17(5884)

General description

RFC-like DNA damage sensor (RAD17)/checkpoint clamp loader component, a homolog of the Schizosaccharomyces pombe checkpoint gene, rad17 is located on human chromosome 5q13.2. This gene is abundantly expressed in the testis.

Specificity

Anti RAD17 (C-terminal) specifically recognizes RAD17 (70 kDa and 75 kDa).

Immunogen

synthetic peptide corresponding to amino acids 623-640 of human Rad17 (isoform 2), conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide is present in isoforms 1-4 of RAD17.

Application

Anti-RAD17 (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

RFC-like DNA damage sensor (RAD17)/checkpoint clamp loader component participates in DNA replication, repair and modification or chromatin remodeling. Phosphorylation of RAD17 is a critical event during checkpoint signaling in DNA-damaged cells. RAD17 has been reported in colon, breast and non-small cell lung carcinoma. It may also participate in cell cycle control.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Resolution Genome Wide Copy Number Alteration (CNA) and Loss of Heterozigosity (LOH) Analysis in Chronic Myeloid Leukemia (CML) Shows That High and Intermediate Sokal Risk Pts (Pts) Have Multiple Losses Targeting Genes Involved in DNA Repair.
Soverini S, et al.
Blood, 114(22), 3262-3262 (2009)
HRad17, a human homologue of the Schizosaccharomyces pombe checkpoint gene rad17, is overexpressed in colon carcinoma
Bao S, et al.
Cancer research, 59(9), 2023-2028 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service