R5880
Renin Substrate Tetradecapeptide human
≥95% (HPLC)
Synonym(s):
Angiotensinogen 1-14, Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Asn
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C83H122N24O19
CAS Number:
Molecular Weight:
1760.01
MDL number:
UNSPSC Code:
12352200
Assay
≥95% (HPLC)
storage temp.
−20°C
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M D Mostaqul Huq et al.
Journal of proteome research, 8(3), 1156-1167 (2009-02-17)
Receptor interacting protein 140 (RIP140) undergoes extensive post-translational modifications (PTMs), including phosphorylation, acetylation, arginine methylation, and pyridoxylation. PTMs affect its subcellular distribution, protein-protein interaction, and biological activity in adipocyte differentiation. Arginine methylation on Arg(240), Arg(650), and Arg(948) suppresses the repressive
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service