Skip to Content
Merck
All Photos(1)

Key Documents

R5509

Sigma-Aldrich

Nde I from Neisseria denitrificans

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-CA/TATG-3′
Cutting results: a 2-10-fold Nde I overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: Inactivated at 60 °C for 15 minutes.

Application

NdeI is a restriction endonuclease used in molecular biology methods to cleave DNA at the recognition site 5′-CA/TATG-3′, generating fragments with 5′-cohesive ends.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SH (B3657).

Unit Definition

One unit is the enzyme activity that completely cleaves 1 mg λDNA in 1 hr. at 37 °C in a total volume of 25 mL of restriction endonuclease buffer SH.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 0.1 mM EDTA, 100 mM NaCl, 1 mM dithioerythritol, 0.02% polydocanol, 0.01% gelatine, 50% glycerol (v/v) at 4 °C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vega Masignani et al.
The Journal of experimental medicine, 197(6), 789-799 (2003-03-19)
Sepsis and meningitis caused by serogroup B meningococcus are devastating diseases of infants and young adults, which cannot yet be prevented by vaccination. By genome mining, we discovered GNA1870, a new surface-exposed lipoprotein of Neisseria meningitidis that induces high levels
NdeI: a restriction endonuclease from Neisseria denitrificans which cleaves DNA at 5'-CATATG-3' sequences.
R J Watson et al.
FEBS letters, 150(1), 114-116 (1982-12-13)
Cynthia L Richard-Fogal et al.
Journal of bacteriology, 190(10), 3489-3493 (2008-03-11)
The system I cytochrome c biogenesis pathway requires CcmD, a small polypeptide of 69 residues in Escherichia coli. Here it is shown that CcmD is a component of the CcmABC ATP-binding cassette transporter complex. CcmD is not necessary for the
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Nico Mitro et al.
Methods in molecular biology (Clifton, N.J.), 952, 137-144 (2012-10-27)
The role of certain amino acids in the interactions of ligands with their cognate nuclear receptors is usually achieved by the resolution of the crystal structure of the receptor complexed with the ligand. As a complementary functional approach, site-directed mutagenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service