Skip to Content
Merck
All Photos(1)

Key Documents

R5258

Sigma-Aldrich

Riboflavin Binding Protein from chicken egg white

lyophilized powder

Synonym(s):

Binding Protein, Chicken Egg White Protein, Riboflavin Protein

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€66.70
10 MG
€189.00

€66.70


Estimated to ship on11 May 2025



Select a Size

Change View
2 MG
€66.70
10 MG
€189.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€66.70


Estimated to ship on11 May 2025


biological source

chicken egg white

Quality Level

form

lyophilized powder

concentration

≥50% protein (biuret)

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... RBP(396449)

General description

Riboflavin Binding Protein (RBP) comprises of 19 cysteines and corresponds to a molecular weight of 30 kDa.[1] Egg white RBP is produced in the oviduct cells. Chicken egg white RBP contains an N-terminal ligand binding domain and a C-terminal phosphorylated motif.[2]
Riboflavin binding protein (RBP) transports riboflavin through the bloodstream of the laying hen into the eggs, where the vitamin is essential for embryonic growth and development. RBP consists of a single polypeptide chain containing 9 disulfide bonds. It is a glycoprotein containing ~14% carbohydrate, with sugar moieties including galactose, mannose and glucosamine, and is phosphorylated.

Application

Riboflavin Binding Protein from chicken egg white may be used in fluorescence quenching experiments.[1]

Biochem/physiol Actions

Riboflavin Binding Protein (RBP) is crucial for normal embryo development.[1] RBP interacts with riboflavin and its analogs. The RBP-riboflavin complex is stabilized by interaction between the isoalloxazyne ring of riboflavin and the tyrosine 75 and tryptophan 156 of residues of RBP.[3] RBP elicits sweet supressing functionality and may interact with the sweet taste receptor.[4]
Riboflavin binding protein binds to flavokinase and regulates its activity. [5]

Preparation Note

Supplied as the riboflavin-depleted apo-form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riboflavin-binding protein exhibits selective sweet suppression toward protein sweeteners
Maehashi K, et al.
Chemical Senses, 32(2), 183-190 (2006)
Binding study of riboflavin-binding protein with riboflavin and its analogues by differential scanning calorimetry
Wasylewski M
Journal of Protein Chemistry, 19(6), 523-528 (2000)
Crystal structure of chicken riboflavin-binding protein
Monaco HL
The Embo Journal, 16(7), 1475-1483 (1997)
Electrochemistry of riboflavin-binding protein and its interaction with riboflavin
Bartovsik M, et al.
Bioelectrochemistry, 76(1-2), 70-75 (2009)
The effect of riboflavin binding protein (RPB) on flavokinase catalytic activity.
Slomczynska, M.
Comparative Biochemistry and Physiology, 87, 681-685 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service