Skip to Content
Merck
All Photos(3)

Key Documents

R3655

Sigma-Aldrich

Rubitecan

powder, ≥98% (HPLC)

Synonym(s):

9-nitrocamptothecin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€54.10
50 MG
€223.00

€54.10


Check Cart for Availability


Select a Size

Change View
10 MG
€54.10
50 MG
€223.00

About This Item

Empirical Formula (Hill Notation):
C20H15N3O6
CAS Number:
Molecular Weight:
393.35
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

€54.10


Check Cart for Availability

Product Name

Rubitecan,

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

mp

182-186 °C

solubility

DMSO: 1 mg/mL

storage temp.

room temp

SMILES string

CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(cccc5nc34)[N+]([O-])=O)C2=O

InChI

1S/C20H15N3O6/c1-2-20(26)13-7-16-17-10(8-22(16)18(24)12(13)9-29-19(20)25)6-11-14(21-17)4-3-5-15(11)23(27)28/h3-7,26H,2,8-9H2,1H3/t20-/m0/s1

InChI key

VHXNKPBCCMUMSW-FQEVSTJZSA-N

Biochem/physiol Actions

Topoisomerase I inhibitor. Rubitecan induces protein- linked DNA single strand breaks, blocking DNA and RNA synthesis in dividing cells. This mode of action makes rubitecan a potential chemotherapeutic agent, and it has been used with some success against refractory pancreatic cancer.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunzhen Zheng et al.
PloS one, 6(6), e21064-e21064 (2011-06-23)
Hepatocellular carcinoma (HCC) is the third most common cause of cancer related mortality worldwide. 9-Nitrocamptothecin (9NC) is a potent topoisomerase-I inhibitor with strong anticancer effect. To increase the solubility and stability, we synthesized a novel 9NC loaded liposomes (9NC-LP) via
Katayoun Derakhshandeh et al.
International journal of nanomedicine, 5, 463-471 (2010-10-20)
The purpose in this study was to investigate poly(ethylene glycol)-modified poly (d,l-lactide-co-glycolide) nanoparticles (PLGA-PEG-NPs) loading 9-nitrocamptothecin (9-NC) as a potent anticancer drug. 9-NC is an analog of the natural plant alkaloid camptothecin that has shown high antitumor activity and is
J M Gao et al.
Biomedical materials (Bristol, England), 3(1), 015013-015013 (2008-05-07)
9-nitro-20(S)-camptothecin (9-NC) is a potent topoisomerase-I inhibitor, and it was applied for clinical trials in cancer treatment. However, the applications of 9-NC were limited by its poor solubility and instability. In order to overcome these disadvantages, 9-NC was encapsulated in
Alfonso Quintas-Cardama et al.
Cancer, 107(7), 1525-1529 (2006-09-07)
Topoisomerase I inhibitors, like topotecan, have activity in myelodysplastic syndrome (MDS) and chronic myelomonocytic leukemia (CMML). 9-Nitro-camptothecin (9-NC) is a new oral topoisomerase inhibitor with a good safety profile. The aims of the current study were to evaluate the activity
Ye Jiang et al.
International journal of pharmaceutics, 397(1-2), 116-121 (2010-07-20)
The effect of a series of cyclodextrins (CDs), especially hydroxypropyl-beta-cyclodextrin (HP-beta-CD), on aqueous solubility and chemical stability of 9-nitro-camptothecin (9-NC), was investigated with an aim of preparing a stable and effective parenteral formulation. The 9-NC/HP-beta-CD complex was obtained in solid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service