Skip to Content
Merck
All Photos(1)

Key Documents

R2507

Sigma-Aldrich

Reactive Blue 4−Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.56

form

saline suspension

extent of labeling

2-7 mg per mL

matrix

cross-linked 4% beaded agarose

capacity

≥5 mg/mL binding capacity (human serum albumin)

storage temp.

2-8°C

SMILES string

O=C1C2=C(C(NC3=CC=C(S(=O)(O)=O)C(NC4=NC(Cl)=NC(Cl)=N4)=C3)=CC(S(=O)(O)=O)=C2N)C(C5=C1C=CC=C5)=O

InChI

1S/C23H14Cl2N6O8S2/c24-21-29-22(25)31-23(30-21)28-12-7-9(5-6-14(12)40(34,35)36)27-13-8-15(41(37,38)39)18(26)17-16(13)19(32)10-3-1-2-4-11(10)20(17)33/h1-8,27H,26H2,(H,34,35,36)(H,37,38,39)(H,28,29,30,31)

InChI key

RTLULCVBFCRQKI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Reactive Blue 4-agarose is used in affinity chromatography, protein chromatography and dye resins. Reactive Blue 4-agarose has been used to express human pituitary glutaminyl cyclase (hQC) in Drosophila S2 cells.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Saeed, S.A., et al.
J. Chem. Soc. Pak., 29(4), 364-372 (2007)
F L Brinkhaus et al.
Archives of biochemistry and biophysics, 266(2), 607-612 (1988-11-01)
Geranylgeranyl diphosphate synthase has been purified to homogeneity from the carotene-overproducing strain M1 of Phycomyces blakesleanus. Usually two activity peaks with molecular weights of 60,000 and 30,000 eluted on gel exclusion chromatography, suggesting that the enzyme consists of two subunits
S Roychoudhury et al.
The Journal of biological chemistry, 269(16), 12067-12073 (1994-04-22)
Intrinsic resistance toward beta-lactams in methicillin-resistant Staphylococcus aureus strains, a major source of nosocomial infections, is believed to be caused mainly by penicillin-binding protein 2a (PBP2a). This protein resembles other penicillin-binding proteins that are involved in bacterial cell wall biosynthesis
Matthew C T Hartman et al.
Biochemistry, 46(41), 11630-11638 (2007-09-22)
In glycosyltransferase-catalyzed reactions a new carbohydrate-carbohydrate bond is formed between a carbohydrate acceptor and the carbohydrate moiety of either a sugar nucleotide donor or lipid-linked saccharide donor. It is currently believed that most glycosyltransferase-catalyzed reactions occur via an electrophilic activation
H Takeshita et al.
FEBS letters, 440(1-2), 239-242 (1998-12-23)
We purified DNase II from human liver to apparent homogeneity. The N-terminal amino acid sequences of each of three components constituting the purified mature enzyme were then separately determined by automatic Edman degradation. A combination of this chemical information and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service