Skip to Content
Merck
All Photos(1)

Key Documents

R1406

Sigma-Aldrich

RIPK2 (1-299), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CARD3, CARDIAK, RICK, RIP2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€495.00

€495.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€495.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

€495.00


Estimated to ship on28 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17-23 nmol/min·mg

mol wt

~59 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RIPK2(8767)

General description

Receptor interacting serine/threonine kinase 2 (RIPK2) or RICK is a serine/threonine kinase.[1] It contains a kinase domain at the amino-terminal, an intermediate domain and a caspase activation and recruitment domain (CARD) at the carboxy-terminal.[2] The gene encoding RIPK2 is localized on human chromosome 8q21.3.[3]

Biochem/physiol Actions

Receptor interacting serine/threonine kinase 2 (RIPK2) or RICK is activated during innate immune responses and acts as a scaffold for downstream effectors.[4] It functions in nuclear factor-κB (NFκB) activation pathways.[1] The protein also has roles in toll-like receptor (TLR)-signaling pathways and in the production of inflammatory cytokines.[5]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of receptor-interacting serine/threonine protein kinase-2 (RIPK2) affects EMT-associated gene expression in human hepatoma cells.
Wu S
Anticancer Research, 3775-3783 (2012)
Structures of the inactive and active states of RIP2 kinase inform on the mechanism of activation.
Pellegrini E
PLoS ONE (2017)
N Inohara et al.
The Journal of biological chemistry, 273(20), 12296-12300 (1998-06-20)
Signaling through the CD95/Fas/APO-1 death receptor plays a critical role in the homeostasis of the immune system. RICK, a novel protein kinase that regulates CD95-mediated apoptosis was identified and characterized. RICK is composed of an N-terminal serine-threonine kinase catalytic domain
Primary urethral clear-cell adenocarcinoma: comprehensive analysis by surgical pathology, cytopathology, and next-generation sequencing.
Rohit Mehra
The American Journal of Pathology (2014)
RIP2, a checkpoint in myogenic differentiation.
Munz B
Molecular and Cellular Biology (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service