Skip to Content
Merck
All Photos(1)

Key Documents

R1283

Sigma-Aldrich

Rhodblock 6

≥98% (HPLC)

Synonym(s):

N-1H-Indazol-5-yl-cyclobutanecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€113.00
25 MG
€451.00

€113.00


Check Cart for Availability


Select a Size

Change View
5 MG
€113.00
25 MG
€451.00

About This Item

Empirical Formula (Hill Notation):
C12H13N3O
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€113.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

faintly bluish
off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

O=C(Nc1ccc2[nH]ncc2c1)C3CCC3

InChI

1S/C12H13N3O/c16-12(8-2-1-3-8)14-10-4-5-11-9(6-10)7-13-15-11/h4-8H,1-3H2,(H,13,15)(H,14,16)

InChI key

GXJXOQKXBIPBKB-UHFFFAOYSA-N

Biochem/physiol Actions

Rhodblock 6 is an inhibitor of the Rho Kinase pathway.
Rhodblock 6 is an inhibitor of the Rho Kinase pathway. Rhodblock 6 directly inhibited Rok and its human ortholog ROCK I in a kinase assay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xavier Fioramonti et al.
Diabetes, 59(2), 519-528 (2009-11-26)
The response of ventromedial hypothalamic (VMH) glucose-inhibited neurons to decreased glucose is impaired under conditions where the counterregulatory response (CRR) to hypoglycemia is impaired (e.g., recurrent hypoglycemia). This suggests a role for glucose-inhibited neurons in the CRR. We recently showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service