Skip to Content
Merck
All Photos(1)

Key Documents

R1008

Sigma-Aldrich

Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse

clone RG-96, ascites fluid

Synonym(s):

Monoclonal Anti-Rabbit IgG (γ-chain specific)

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
€173.00
0.5 ML
€761.00

€173.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
0.2 ML
€173.00
0.5 ML
€761.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€173.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

RG-96, monoclonal

contains

15 mM sodium azide

species reactivity

rabbit

should not react with

goat, feline, pig, guinea pig, rat, bovine, canine, human, horse, sheep, chicken

technique(s)

dot blot: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable
indirect ELISA: 1:2,000
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Monoclonal Anti-Rabbit IgG (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulins (Igs) belongs immunoglobulin super-family. There are five different types of immunoglobulins. Each immunoglobulin has two heavy (H) and two light (L), held together by disulphide linkages. Each light comprises one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region.
The product recognizes an epitope located on the γ (heavy)-chain of rabbit IgG. In immunoblotting, the antibody recognizes both native and denatured forms of rabbit IgG. In ELISA, the antibody is specific for rabbit IgG, and shows no cross-reactivity with rabbit IgA and IgM or human IgG, IgA, and IgM. No cross reaction is observed with IgG from the following species: bovine, cat, chicken, dog, goat, guinea pig, horse, pig, rat, or sheep.

Specificity

Monoclonal Anti-Rabbit IgG is specific for an epitope located on the heavy (γ) chain of rabbit IgG, in an immunoblot of denatured, reduced rabbit IgG. The antibody does not react with rabbit IgA, IgM or light chains. No cross reactivity is observed with IgG from bovine, cat, chicken, dog, goat, guinea pig, horse, human, pig, rat or sheep.

Application

Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse has been used in
  • enzyme linked immunosorbent assay (ELISA)
  • dot blots
  • immunohistology
  • western blotting
  • peptide mass fingerprinting (PMF)

Biochem/physiol Actions

Digestion of IgG by papain generates fragment antigen binding (Fab). Pepsin digestion of IgG results in fragment crystallisable (fc). IgG antibody has enormous therapeutic potential and the Fc region is involved in the development of therapeutic antibody.

Physical form

The product is provided as ascites fluid with 15 mM sodium azide (see MSDS) as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy K Brown et al.
The American journal of pathology, 165(1), 95-106 (2004-06-25)
Peak intestinal mucosal mast cell (MMC) recruitment coincides with expulsion of Trichinella spiralis, at a time when the majority of the MMCs are located within the epithelium in BALB/c mice. Although expression of integrin-alpha(E)beta(7) by MMCs has not been formally
Specificity in killing pathogens is mediated by distinct repertoires of human neutrophil peptides
Cederlund A, et al.
Journal of Innate Immunity, 2(6), 508-521 (2010)
Andreas Cederlund et al.
Journal of innate immunity, 2(6), 508-521 (2010-09-08)
Neutrophil-derived antimicrobial peptides and proteins (AMPs) play an important role in the defense against microbes. Absence of defense is illustrated by neutropenic patients with frequent bacterial and fungal infections. However, the specificity of the antimicrobial effects has not been adequately
Rossalyn M Day et al.
Cell stress & chaperones, 8(2), 183-193 (2003-11-25)
Small heat shock/alpha-crystallin proteins function as molecular chaperones, protecting other proteins from irreversible denaturation by an energy-independent process. The brine shrimp, Artemia franciscana, produces a small heat shock/alpha-crystallin protein termed p26, found in embryos undergoing encystment, diapause, and metabolic arrest.
Presenilins are enriched in endoplasmic reticulum membranes associated with mitochondria
Area-Gomez E, et al.
The American Journal of Pathology, 175(5), 1810-1816 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service