Skip to Content
Merck
All Photos(1)

Key Documents

R0783

Sigma-Aldrich

RG-239 hydrate

Synonym(s):

(3)-3-Hydroxy-3-(2-propen-1-yl)-lup-20(29)-en-28-oic acid hydrate, (3R)-3-Allyl-3-hydroxylup-20(29)-en-28-oic acid hydrate, 18 dia 2

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€144.00

€144.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
€144.00

About This Item

Empirical Formula (Hill Notation):
C33H52O3 · xH2O
Molecular Weight:
496.76 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€144.00


Estimated to ship on29 May 2025


Assay

≥98% (HPLC)

form

solid

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O.CC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@@](O)(CC=C)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(O)=O

InChI

1S/C33H52O3.H2O/c1-9-14-33(36)20-17-29(6)24(28(33,4)5)13-15-31(8)25(29)11-10-23-26-22(21(2)3)12-16-32(26,27(34)35)19-18-30(23,31)7;/h9,22-26,36H,1-2,10-20H2,3-8H3,(H,34,35);1H2/t22-,23+,24-,25+,26+,29-,30+,31+,32-,33-;/m0./s1

InChI key

JMELBGCTIDKOPM-RQBNYOBGSA-N

Application

RG-239, a betulinic acid derivative and TGR5/Gpbar-1 agonist, is used to study the role of bile acid activated TGR5 in intestinal motility, inflammation, energy production, glucose homeostasis, glycemic control, obesity and liver steatosis.

Biochem/physiol Actions

RG-239 is a TGR5 agonist
Semisynthetic derivative of betulinic acid (3β-allylbetulinic acid), RG-239 is an agonist at the G-protein coupled receptor TGR5. TGR5 signalling induces glucagon-like peptide-1 (GLP-1) release in brown adipose tissue, enhancing glucose tolerance and attenuating diet-induced obesity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service