Skip to Content
Merck
All Photos(1)

Key Documents

R0758

Sigma-Aldrich

Ritodrine hydrochloride

Synonym(s):

N-(p-Hydroxyphenethyl)-4-hydroxynorephedrine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€121.00
1 G
€403.00

€121.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€121.00
1 G
€403.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO3 · HCl
CAS Number:
Molecular Weight:
323.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€121.00


Estimated to ship on28 May 2025


Request a Bulk Order

originator

AstraZeneca

SMILES string

Cl.C[C@H](NCCc1ccc(O)cc1)[C@H](O)c2ccc(O)cc2

InChI

1S/C17H21NO3.ClH/c1-12(17(21)14-4-8-16(20)9-5-14)18-11-10-13-2-6-15(19)7-3-13;/h2-9,12,17-21H,10-11H2,1H3;1H/t12-,17-;/m0./s1

InChI key

IDLSITKDRVDKRV-XHXSRVRCSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

β2-adrenoceptor agonist; relaxes uterine muscle contraction.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kimura et al.
Endocrinology, 140(2), 722-731 (1999-02-02)
The regulation of mitogen-activated protein (MAP) kinase by endothelin-1 (ET-1) in cultured rat puerperal uterine myometrial cells was investigated. ET-1 caused the rapid stimulation of MAP kinase activity. ET-1-induced MAP kinase activation is neither extracellular Ca2+- nor intracellular Ca2+-dependent. ET-1
J M Bassett et al.
The American journal of physiology, 275(1 Pt 2), R112-R119 (1998-08-05)
Prolonged administration of the beta2-adrenergic agonist ritodrine to fetal sheep increases nonesterified fatty acid mobilization. To investigate whether changes in fetal growth or functional development of brown adipose tissue (BAT) also occur, ritodrine was infused at 5 microg/min iv into
S Bertini et al.
General pharmacology, 31(4), 625-631 (1998-10-29)
1. The involvement of beta3 adrenoceptors in the control of gastric acid secretion and histamine release was investigated in the dog. 2. In conscious dogs, SR 58611A (0.0625-1.0 mg/kg/hr i.v.) dose dependently inhibited gastric acid secretion induced by pentagastrin. Maximal
Hiraku Onishi et al.
International journal of pharmaceutics, 468(1-2), 207-213 (2014-04-09)
Ritodrine hydrochloride (RD-HCl) tablets containing alginate (AL) and lactose (LC) with or without microcrystalline cellulose (MC) as excipients were produced as a buccal dosage form. The RD-HCl (2 mg) tablets with AL/LC but no MC swelled and dissolved gradually in
Simona Codruţa Cobzac et al.
Journal of chromatographic science, 52(9), 1095-1103 (2013-10-29)
The retention behavior for a series of amine neurotransmitters, their precursors, metabolites and structurally related drugs has been investigated in reversed-phase thin-layer chromatography using RP-18, RP-8, RP-2, CN and Diol stationary phases and mixtures of phosphate buffer and methanol as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service