Skip to Content
Merck
All Photos(1)

Key Documents

PZ0357

Sigma-Aldrich

PF-06412562

≥98% (HPLC)

Synonym(s):

4-[4-(4,6-Dimethyl-5-pyrimidinyl)-3-methylphenoxy]-1H-pyrazolo[4,3-c]pyridine, 4-[4-(4,6-Dimethylpyrimidin-5-yl)-3-methylphenoxy]-1H-pyrazolo[4,3-c]pyridine, PF 06412562, PF06412562

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€79.30
25 MG
€347.00

€79.30


Check Cart for Availability


Select a Size

Change View
5 MG
€79.30
25 MG
€347.00

About This Item

Empirical Formula (Hill Notation):
C19H17N5O
CAS Number:
Molecular Weight:
331.37
UNSPSC Code:
41105101
NACRES:
NA.77

€79.30


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=C(C2=C(C)N=CN=C2C)C=CC(OC3=NC=CC4=C3C=NN4)=C1

InChI

1S/C19H17N5O/c1-11-8-14(25-19-16-9-23-24-17(16)6-7-20-19)4-5-15(11)18-12(2)21-10-22-13(18)3/h4-10H,1-3H3,(H,23,24)

InChI key

IDIUJOHYYBNCPC-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06412562 is a dopamine receptor D1 (DRD1) agonist ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service