Skip to Content
Merck
All Photos(1)

Key Documents

PZ0347

Sigma-Aldrich

CP-424,174

≥98% (HPLC)

Synonym(s):

CP 424174, CP-424174, CP424174, N-[[[4-Chloro-2,6-bis(1-methylethyl)phenyl]amino]carbonyl]-3-(1-hydroxy-1-methylethyl)-benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€91.80
25 MG
€374.00

€91.80


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
€91.80
25 MG
€374.00

About This Item

Empirical Formula (Hill Notation):
C22H29ClN2O4S
CAS Number:
Molecular Weight:
452.99
UNSPSC Code:
12352200
NACRES:
NA.77

€91.80


Estimated to ship on31 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(O)(C)C1=CC(S(=O)(NC(NC2=C(C(C)C)C=C(Cl)C=C2C(C)C)=O)=O)=CC=C1

InChI

1S/C22H29ClN2O4S/c1-13(2)18-11-16(23)12-19(14(3)4)20(18)24-21(26)25-30(28,29)17-9-7-8-15(10-17)22(5,6)27/h7-14,27H,1-6H3,(H2,24,25,26)

InChI key

YRSBLSHMKVQWHP-UHFFFAOYSA-N

Biochem/physiol Actions

CP-424,174 is a cytokine release inhibitory drugs (CRID) that inhibits the post-translational processing and secretion of IL-1b in response to LPS and ATP in human monocytes. It is quite possible that similarly to CP-456,773 (termed CRID3), CP-424,174 inhibits both NLRP3 and AIM2 inflammasomes by preventing ASC oligomerisation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David G Perregaux et al.
Journal of immunology (Baltimore, Md. : 1950), 168(6), 3024-3032 (2002-03-09)
Human monocytes stimulated with LPS produce large quantities of prointerleukin-1beta, but little of this cytokine product is released extracellularly as the mature biologically active species. To demonstrate efficient proteolytic cleavage and export, cytokine-producing cells require a secondary effector stimulus. In
D G Perregaux et al.
The Journal of pharmacology and experimental therapeutics, 299(1), 187-197 (2001-09-19)
Lipopolysaccharide (LPS)-activated monocytes and macrophages produce large quantities of pro-interleukin (IL)-1beta but externalize little mature cytokine. Efficient post-translational processing of the procytokine occurs in vitro when these cells encounter a secretion stimulus such as ATP, cytolytic T cells, or hypotonic
Rebecca C Coll et al.
PloS one, 6(12), e29539-e29539 (2012-01-05)
The Inflammasomes are multi-protein complexes that regulate caspase-1 activation and the production of the pro-inflammatory cytokine IL-1β. Previous studies identified a class of diarylsulfonylurea containing compounds called Cytokine Release Inhibitory Drugs (CRIDs) that inhibited the post-translational processing of IL-1β. Further

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service