Skip to Content
Merck
All Photos(1)

Key Documents

PZ0299

Sigma-Aldrich

PF-05175157

≥98% (HPLC)

Synonym(s):

1,4-Dihydro-1′-[(2-methyl-1H-benzimidazol-6-yl)carbonyl]-1-(1-methylethyl)-spiro[5H-indazole-5,4′-piperidin]-7(6H)-one, 1-Isopropyl-1-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-1,4-dihydrospiro[indazole-5,4-piperidin]-7(6H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€128.00
25 MG
€517.00

€128.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€128.00
25 MG
€517.00

About This Item

Empirical Formula (Hill Notation):
C23H27N5O2
CAS Number:
Molecular Weight:
405.49
UNSPSC Code:
12352200
NACRES:
NA.77

€128.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC(C)N(N=C1)C2=C1CC3(CCN(C(C4=CC(NC(C)=N5)=C5C=C4)=O)CC3)CC2=O

InChI

1S/C23H27N5O2/c1-14(2)28-21-17(13-24-28)11-23(12-20(21)29)6-8-27(9-7-23)22(30)16-4-5-18-19(10-16)26-15(3)25-18/h4-5,10,13-14H,6-9,11-12H2,1-3H3,(H,25,26)

InChI key

BDXXSFOJPYSYOC-UHFFFAOYSA-N

Application

PF-05175157 has been used as an acetyl-Coenzyme A carboxylase (ACC) inhibitor to study its effects on the infection of flavivirus.[1]

Biochem/physiol Actions

PF-05175157 is a potent and selective inhibitor of both acetyl-CoA carboxylase isoform ACC1 located primarily in liver and adipose tissue and isoform ACC2 dominant in skeletal and heart muscle, with IC50 values of 27 nM and 33 nM, respectively. Acetyl CoA carboxylase (ACC) generates malonyl CoA, which is a substrate for de novo lipogenesis and is also an inhibitor of mitochondrial fatty acid β-oxidation through inihbition of carnitine-palmitoyl transferase I (CPT-1), responsible for the transport of long-chain fatty acyl-CoAs across the mitochondrial membrane. ACC inihibitors are hoped to inhibit de novo lipogenesis and increase β-oxidation of long-chain fatty acids with potential for treatment of type 2 diabetes, hepatic steatosis, and cancer. In Phase I clinical studies for diabetes treatment, PF-05175157 inhibited de novo lipogenesis and increased net whole-body fatty acid utilization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nereida Jiménez de Oya et al.
Emerging microbes & infections, 8(1), 624-636 (2019-04-20)
Flaviviruses are (re)-emerging RNA viruses strictly dependent on lipid metabolism for infection. In the search for host targeting antivirals, we explored the effect of pharmacological modulation of fatty acid metabolism during flavivirus infection. Considering the central role of acetyl-Coenzyme A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service