PZ0292
PF-06658959-00
≥98% (HPLC)
Synonym(s):
4-(4-Chloro-3-methoxy-phenylamino)-6-methoxy-7-[3-(4-methyl-piperazin-1-yl)-propoxy]-quinoline-3-carbonitrile, Bosutinib analog 7, PF-06658959
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H30ClN5O3
Molecular Weight:
496.00
UNSPSC Code:
12352200
NACRES:
NA.77
Assay
≥98% (HPLC)
form
powder
color
white to light brown
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
room temp
Biochem/physiol Actions
PF-05883083-00 is an analog of orally available dual Src/Abl kinase inhibitor Bosutinib.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service