Skip to Content
Merck
All Photos(1)

Key Documents

PZ0277

Sigma-Aldrich

PF-06281355

≥98% (HPLC)

Synonym(s):

6-(2,5-Dimethoxyphenyl)-3,4-dihydro-4-oxo-2-thioxo-1(2H)-pyrimidineacetamide, PF 6281355, PF-1355

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€109.00
25 MG
€351.00

€109.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€109.00
25 MG
€351.00

About This Item

Empirical Formula (Hill Notation):
C14H15N3O4S
CAS Number:
Molecular Weight:
321.35
UNSPSC Code:
51111800
NACRES:
NA.77

€109.00


Estimated to ship on13 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

room temp

Biochem/physiol Actions

PF-06281355 (PF-1355) is an orally available, selective and potent mechanism based inhibitor of the myeloperoxidase (MPO) that reduces plasma MPO activity, vascular edema, neutrophil recruitment, and elevated circulating cytokines. PF-06281355 suppresses albuminuria and chronic renal dysfunction in model of anti-Glomerular Basement Membrane (GBM) disease.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service