Skip to Content
Merck
All Photos(1)

Key Documents

PZ0264

Sigma-Aldrich

CP-547632 hydrochloride

≥98% (HPLC)

Synonym(s):

3-[(4-Bromo-2,6-difluorophenyl)methoxy]-5-[[[[4-(1-pyrrolidinyl)butyl]amino]carbonyl]amino]-4-Isothiazolecarboxamide hydrochloride, PF-01371182 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€75.80
25 MG
€303.00

€75.80


Estimated to ship on31 March 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€75.80
25 MG
€303.00

About This Item

Empirical Formula (Hill Notation):
C20H24BrF2N5O3S · HCl
CAS Number:
Molecular Weight:
568.86
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€75.80


Estimated to ship on31 March 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

NC(C1=C(NC(NCCCCN2CCCC2)=O)SN=C1OCC3=C(F)C=C(Br)C=C3F)=O.[H]Cl

InChI

1S/C20H24BrF2N5O3S.ClH/c21-12-9-14(22)13(15(23)10-12)11-31-18-16(17(24)29)19(32-27-18)26-20(30)25-5-1-2-6-28-7-3-4-8-28;/h9-10H,1-8,11H2,(H2,24,29)(H2,25,26,30);1H

InChI key

NNPJZJJMQDXYGN-UHFFFAOYSA-N

Biochem/physiol Actions

CP-547632 hydrochloride is a well-tolerated, orally-bioavailable ATP-competitive kinase inhibitor. It is currently in clinical trials for the treatment of human malignancies.
CP-547632 is a potent inhibitor of the VEGFR-2 and basic fibroblast growth factor (FGF) kinases (IC50 = 11 and 9 nM, respectively). The compound CP-547632 inhibits VEGF and basic FGF-induced angiogenesis in animal models, and inhibits growth of xenograft tumors in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean S Beebe et al.
Cancer research, 63(21), 7301-7309 (2003-11-13)
Signaling through vascular endothelial growth factor (VEGF) receptors (VEGFRs) is a key pathway initiating endothelial cell proliferation and migration resulting in angiogenesis, a requirement for human tumor growth and metastasis. Abrogation of signaling through VEGFR by a variety of approaches

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service